Would you like to log in again?
Optical Lens Cleaner, Aldon
Supplier: Aldon
LL0158-8OZ
470153-310EA 10 USD InStock
470153-310
Optical Lens Cleaner, Aldon
Cleaning Agents Lens Cleaning Solutions
Cleans without leaving residue.
  • Store between 40 to 100 °F
Our concentrated formula cleans glass or hard plastic safely, quickly and effectively. Simply apply and wipe clean. Unlike similar fluids, it cleans without leaving a residue, does not exhibit an annoying odor, and does not dry too quickly, making it ideal for precision lens, window, or prism cleaning.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty