Would you like to log in again?
Porcelain Buchner Funnels
Supplier: SILICO & CHEMICO PORCELAIN SE
J-187 J-285 J-283 J-284
470149-558CS 153 USD InStock
470149-558 470149-206 470153-508 470149-039
Porcelain Buchner Funnels
Funnels Filtration Funnels Büchner Funnels
High Quality yet Affordable Buchner Funnels.
  • Two sizes available
  • Fragile
These high quality Buchner funnels are made from glazed perforated plate high-quality porcelain. They have a fixed, perforated plate and are glazed on the inside and outside with exception of the rim.
Caution: Fragile, use care when handling.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty