Would you like to log in again?
Polyethylene Disposable Gloves
Supplier: DIVAL SAFETY & SUPPLIES SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
GDPEMED GDPE-LG GDPEMD
470153-722BX 11.95 USD InStock
470153-722 470153-724 470016-978
Polyethylene Disposable Gloves
Gloves
Ultra sensitive.
  • Ambidextrous
  • Powder-Free
These disposable, polyethylene gloves feature sheer-gauge 1.25 mil, which permits near bare hand sense of touch while protecting hands and labware from contamination. Gloves are powder-free, so they will not mark delicate materials.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty