Would you like to log in again?
SP Bel-Art Mercury Collector, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
F36250-0000_SE
470153-828CS 537.8 USD InStock
470153-828
SP Bel-Art Mercury Collector, Bel-Art Products, a part of SP
Spill Control Materials
Simply press the foam collector onto elusive beads of spilled mercury and the foam pores open to enclose it. Closing the container forces the beads into a recessed trap. One replacement pad is included.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty