Would you like to log in again?
Plastic Slide Box for 25 Slides
Supplier: C & A SCIENTIFIC CO INC
97-0025
470156-718CS 1085.55 USD InStock
470156-718
Plastic Slide Box for 25 Slides
Microscope Slide Boxes
Store your slides efficiently.
  • Store your slides safely and keep them well organized
The plastic box protects 1" x 3" slides and won’t warp when the temperature and humidity vary. It is stackable, with telescoping covers, a cork bottom, and embossed slide numbers on the bottom.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty