Would you like to log in again?
Science Take-Out® Atoms, Isotopes And Ions
Supplier: Science Take-Out
STO-142 STO-142U STO-142-NOTG
470157-260EA 96.45 USD InStock
470157-260 470157-259 470157-258
Science Take-Out® Atoms, Isotopes And Ions
Educational Materials Chemistry Educational Materials Ionic Properties Learning Activities
Use Colored Chips To Model Atomic Structure.
  • Available As A Single Kit With Materials For 1 Student Or Group
  • Unassembled 10 Pack For Additional Savings
  • No Additional Equipment Needed
Use colored chips and the information from a periodic table to model the sub-atomic particles (protons, neutrons, and electrons) in atoms, isotopes, and ions. Model the electron configuration within the electron energy levels. Learn how changing the number of protons, neutrons, or electrons leads to different elements, isotopes, or ions. This complete "dry lab" activity contains all required materials.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty