Would you like to log in again?
Science Take-Out® Gene Switches
Supplier: Science Take-Out
STO-143 STO-143-NOTG STO-143U
470157-262EA 15.75 USD InStock
470157-262 470157-264 470157-263
Science Take-Out® Gene Switches
Educational Materials Biology Educational Materials Genetics Learning Activities
How Do Bacteria Turn On And Turn Off Genes?
  • Available As A Single Kit With Materials For 1 Student Or Group
  • Unassembled 10 Pack For Additional Savings
  • No Additional Equipment Needed
Switching genes on and off enables bacteria to conserve resources by producing proteins only when they are needed. Make a model of an operon. Learn how the lac operon regulates the expression of bacterial genes essential for lactose metabolism. This complete "dry lab" activity contains all required materials.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty