Would you like to log in again?
Fundamentals of Physics Photogate System
Supplier: VWR International
PH-14GT PHGST08
470014-466EA 362.45 USD InStock
470014-466 470157-540
Fundamentals of Physics Photogate System
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Versatile System Expands Uses of All FUNdamentals of Physics Products.
  • Easy to navigate timing options
  • Designed to fit onto the Fundamentals of Physics line with no extra equipment
  • External support rod allows use with any other experiment
This innovative photogate system is designed specifically for use with the entire Fundamental of Physics collection. Its unique design can be used for any motion based experiment, and easily attaches to any of the Fundamentals of Physics products. Battery operated, it allows you to conveniently perform experiments without being tethered to an outlet. There are four timing modes, including Gate, which measures the time of interruption; Pulse, which measures the time between interruptions; and Pendulum, which measures the time between the first and third interruption. Includes 9V battery and instructions.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty