Would you like to log in again?
Precision Air Table
Supplier: EISCO SCIENTIFIC LLC
AIRTBL
470157-648EA 790.65 USD InStock
470157-648
Precision Air Table
Educational Materials Physics Educational Materials Mechanics Learning Activities
The air table guides the study of a multitude of concepts.
  • Topics Include Elastic and Inelastic Collisions, Linear and Angular Momentum, and More
  • Air Table Length: 700 mm
  • Width: 500 mm
  • Adjustable Feet
These concepts include elastic and inelastic collisions, linear and angular momentum, conservation of momentum, moment of inertia, centripetal force, and simple and complex harmonic motion.
The air table is 500 x 700 mm with adjustable feet. It requires an air blower.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty