Would you like to log in again?
Linear Air Track Systems
Supplier: EISCO SCIENTIFIC LLC
PH0362A PH0362
470157-664EA 869.4 USD InStock
470157-664 470157-654
Linear Air Track Systems
Educational Materials Physics Educational Materials Mechanics Learning Activities
This kit is designed to provide a cost-effective and friction-free system for the study of all aspects of dynamics.
  • Cost-effective study of dynamics
  • Friction-free environment
  • Effective in the field of collisions
  • Comprehensive accessory pack
  • Other topics include velocity, kinetic energy, and more
It is particularly effective in the field of collisions where conservation of momentum is dramatically demonstrated.
The kit contains everything required for a vast array of experiments covering velocity, force and acceleration, potential energy, kinetic energy, conservation of energy, SHM, and motion on level and inclined planes. Gliders are designed to accept standard slotted masses for additional loads. It is also supplied with a comprehensive accessory pack to facilitate every type of interaction.
Ordering information: This kit includes (1) Linear air track, (1) Accessory pack. Comes with a range of accessories including vehicles with mass attachments, catapults, and buffers. For measurement of glider’s speed, standard light gates and timers are required. This kit does not contain an air blower.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty