Would you like to log in again?
Slide Tray Cabinet
Supplier: BEL-ART PRODUCTS SE
H44581-0000_SE H44583-0000_SE
470201-796EA 402.9 USD InStock
470201-796 470157-670
Slide Tray Cabinet
Microscope Slide Boxes
Store your slides securely with a minimal surface footprint.
  • Store slides horizontally for the best care of whole mounts
  • Clear door allows for internal viewing
Our polypropylene microscope slide tray cabinet holds up to 240 slides total. It can accommodate 12 horizontal slide trays in all. The clear acrylic door slides vertically. Slide trays are available separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty