Would you like to log in again?
Ray Box
Supplier: EISCO SCIENTIFIC LLC
RBX14
470157-710EA 139.75 USD InStock
470157-710
Ray Box
Educational Materials Physics Educational Materials Optics Learning Activities
This Ray Box is a simple, safe, and sturdy source of projected light for many classic optics experiments. Best of all, you don't need a completely dark room to conduct optic experiments due to its high-intensity light!
  • Bright enough to use in ambient light
  • Produces one, three, or five parallel rays
  • May be powered with the included adaptor, or by an external laboratory source
Built in solid aluminum extrusion casing, it is supplied with a multi slit that produces either a single wide beam, or choice of one, three, or five pencil-thin rays. It produces parallel rays across a plane horizontal surface, and the rays are intense enough to be traced in natural light. It is supplied with a power adapter with all international safety certifications, which can be plugged into the mains source 100 to 240 V 50/60 Hz, or an external power supply may be used in the lab. It comes with a fixed lens, a slit with multiple sides, and an adaptor.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty