Would you like to log in again?
Two-Stroke Diesel Engine Model
Supplier: EISCO SCIENTIFIC LLC
PH0490
470157-714EA 134.15 USD InStock
470157-714
Two-Stroke Diesel Engine Model
Educational Materials Physics Educational Materials Mechanics Learning Activities
This model shows the internal structure and operating principles of an air cooled two-stroke engine. All parts are aluminum and plastic. Ignition is shown via a small bulb. Carburetor and fuel supply are also sectioned. Hand crank is provided for manual operation. Mounted on a base.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty