Would you like to log in again?
Superior Jolly Bulb Metal
Supplier: EISCO SCIENTIFIC LLC
PH0138D
470157-716EA 134.4 USD InStock
470157-716
Superior Jolly Bulb Metal
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Visually impacting demonstration of the relationship between temperature and pressure.
  • Easy to read pressure gauge
  • Instruction manual
  • Metal sphere to insure constant volume
A hollow metallic ball 80 mm in diameter is connected to an absolute pressure gauge. Submerging the metal ball in ice water causes pressure to drop, while submerging it in hot water increased the pressure. Utilizing several data points students can calculate the value for absolute zero.
The Superior Jolly Bulb has two scales. The inside scale measures in kPa and the outside scale measures in PSI.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty