Would you like to log in again?
Bugkit
Supplier: LA MOTTE CHEMICAL PROD CO SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5950
470158-074CS 142 USD InStock
470158-074
Bugkit
Educational Materials Biology Educational Materials Ecology Learning Activities
How Healthy is the Stream?
  • Sample insects from a stream
  • Activity can be completed independently
  • includes a teacher's manual
Lead students through a creepy, crawly, wet investigation of the health of streams, ponds, and other freshwater environments. Following the easy-to-read manual, students learn to collect, observe, sort, and identify aquatic insects as a tool for measuring water quality. Students can complete the activity at home after collection or spend an afternoon outdoors beside a stream. Contents include mesh leaf bags for collection, trays and plastic dishes for sorting, a magnifying lens, Bug Guide, and a bug identification chart. A single kit provides materials for one student group.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty