Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Buffer Capsules

Supplier: MICRO ESSENTIAL LABORATORY I

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

270-PH10 270-PH6.0 270-PH7.0 270-PH3.0 270-PH09 270-PH12-B 270-PH8.0 270-PH5.0 270-PH4.0 270-PH11 270-PH2.0
470158-204CS 56.6 USD InStock
470158-204 470158-194 470158-172 470158-182 470158-160 470158-208 470158-176 470158-164 470158-186 470158-168 470158-200
Buffer Capsules
Buffers Cell Culture Buffers
Shelf Life (months): 36
Storage: Green

Caution: Please Note: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty