Would you like to log in again?
Fan Cart
Supplier: United Scientific Supplies
FNCRT1
470158-412EA 36.7 USD InStock
470158-412
Fan Cart
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
This Fan Cart demonstrates Newton’s principle of action and reaction.
  • Fan provides constant force, resulting in constant acceleration
  • Removable plate vividly demonstrates Newton's third law
  • Activity guide included
  • Fan and power unit removable, converting to standard Hall's carriage
  • Propeller thrust 88 mN
The inexpensive fan cart is powered by two AA batteries, not included. The removable power unit contains the batteries, and the fan is controlled by a switch. The propeller delivers a thrust of about 88 mN, accelerating the cart at 0.4 m/s².
A transparent plate can be attached to show the effect of reversing the airflow.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty