Would you like to log in again?
Young's Modules of Wires Apparatus, Eisco Scientific
Supplier: EISCO SCIENTIFIC LLC
PH0328A
470159-556EA 129 USD InStock
470159-556
Young's Modules of Wires Apparatus, Eisco Scientific
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Learn how to measure the stiffness of wires.
  • Aluminum cast frame includes arm with bubble level
  • Micrometer screw
  • Iron weight (approximate): 2.6 lbs.
The Young's modulus of wires apparatus provides all of the necessary tools to study the concept of the measure of the stiffness of a material (Young's Modulus). It provides the tools to study the stress and strain on a material in a visually and kinesthetically effective manner.
Delivery information: Includes aluminum cast frame, micrometer screw, wire holder, two screws, iron weight with hook, and two wires.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty