Would you like to log in again?
Parallel and Series Circuit Demonstration, Eisco Scientific
Supplier: EISCO SCIENTIFIC LLC
PH1128N8-A
470159-582EA 82.25 USD InStock
470159-582
Parallel and Series Circuit Demonstration, Eisco Scientific
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
A simple way to demonstrate parallel and series circuits.
  • Base measures 7.75×4.75"
This parallel and series circuit demonstration can be easily taken apart and reconfigured in a variety of ways to experiment with and observe both parallel and series circuits. This versatile piece of equipment can help students to distinguish between series and parallel circuits and come up with a conceptual and mathematical understanding of how voltage, current, and resistance behave in a series or parallel circuit.
Ordering information: Variable DC power supply (10 V max) required, not included. Includes heavy duty base with 5 bulbs, 8 removable metal bars, and 10 thumb screws.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty