Would you like to log in again?
Magnetizer Box
Supplier: EISCO SCIENTIFIC LLC
MAGBARV1
470159-586EA 258.85 USD InStock
470159-586
Magnetizer Box
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
No need to Buy New Magnets
  • Bar Magnet
  • 110/120V AC
  • 60Hz
Revitalizes iron, nickel, cobalt, and alnico horseshoe and bar magnets in seconds. Simply place magnets in the 25x25 mm holes and press the button to make old magnets new again.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty