Would you like to log in again?
Sherlock Bones: Identification of Skeletal Remains Kit
Supplier: Avantor
COMP-102 WARD470159-672 WARD470190-646 COMP-109
470190-646EA 517.69 USD InStock
470190-646 470030-032 470030-034 470159-672
Sherlock Bones: Identification of Skeletal Remains Kit
Educational Classroom Kits and Activities
Is the victim a male or female?
  • NGSS Life Science DCIs LS1, and LS2.
  • Pre- and post-laboratory assessments
  • Links to a real world scenario
  • Estimated class/lab time required: 45-60 minutes
This lab provides students with experience with the process of epiphyseal union as well as using Vernier calipers. Students will be given a few bones, and then take the role of a forensic anthropologist to determine as much information as possible about the bones to help identify the individual. Students will become familiar with some of the techniques performed by a forensic anthropologist as well as some of the challenges one may encounter along the way. Framework Science Standards (LS2), (LS3), and (LS4) are all utilized in this lab activity (Ecosystems: Interactions, Energy, and Dynamics, Heredity: Inheritance and Variation of Traits, Biological Evolution: Unity and Diversity).
This kit has been aligned with all published National Standards. Pre- and Post-laboratory assessments and vocabulary words all target specific Science and Engineering Practices and common core standards. The teacher guide also contains specific "Did You Know" concepts that link the real world with scientific standards explored. A student copymaster is also included. The lab activity booklet can be used with this kit and our additional bone sets, as well as bone sets that you may already have in your classroom. This kit includes enough materials to be used repeatedly.
Ordering information: Kit contents include Plastic skull, Plastic humerus, Plastic pelvis, Plastic femur, Large caliper, Protractors, Vernier calipers, Metric rulers, Teacher's Guide, and Student Guide. The bones included with this kit were cast from a 25 year old Caucasian male who stood between 5'0" and 5'9" tall. This activity takes approximately 45–60 minutes to complete.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty