Would you like to log in again?
GPI Anatomicals® Basic Hip Model
Supplier: GPI ANATOMICALS
1260
470160-228EA 139.15 USD InStock
470160-228
GPI Anatomicals® Basic Hip Model
Models Human Biology Models
Full size normal right hip with femur includes: articular capsule of the hip joint; greater and lesser trochanter, ilium, ischial spine, ischial tuberosity, ischium and pubis bones.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty