Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

GPI Anatomicals® Canine Shoulder

Supplier: GPI ANATOMICALS
9075
470160-280EA 164.3 USD InStock
470160-280
GPI Anatomicals® Canine Shoulder
Models Zoology Models
Average size shoulder with scapula, humerus, biceps brachii tendon, coracobrachialis tendon, lateral glenohumeral ligament, medial glenohumeral ligament and transverse humeral ligament.

  • Demonstrates Healthy Feline Joints And Arthritis Development
  • Use For Patient Demonstrations, Or Comparative Anatomy
  • Keycard Mounted On Base

Model size: 11-1/8” x 3” x 2”
Card size: 6-1/4” x 8-1/4”
Base size: 6-1/2” x 5”

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty