Would you like to log in again?
GPI Anatomicals® Clear Feline Jaw
Supplier: GPI ANATOMICALS
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
9191
470160-288EA 164.85 USD InStock
470160-288
GPI Anatomicals® Clear Feline Jaw
Models Zoology Models
Examine Feline Dentition Easily With A Clear Model
Healthy feline hinged jaw showing the roots of all teeth (incisors, canine, premolars, molars). Jaws open, close and separate for closer study.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty