Would you like to log in again?
9-Second Flexible Tip Digital Thermometer
Supplier: HEALTH SMART SE
15-736-000
470160-750CS 989 USD InStock
470160-750
9-Second Flexible Tip Digital Thermometer
Thermometers Digital Thermometers
The soft, flexible tip on this waterproof Fahrenheit thermometer conforms to the shape of the mouth for a safe and comfortable reading.
  • Flexible Mouthpiece for More Comfort
  • Large Display Screen
  • Auto Shut off
  • Waterproof
  • Instructions Provided
The easy-to-read, large display screen emits a tone at peak temperature.
Its auto shut off feature saves power.
Ordering information: This kit includes five probe covers, a storage case, instructions, and a lifetime limited manufacturer's warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty