Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Torsion Inertia Device

Supplier: AMERICAN 3B SCIENTIFIC SE
1002836 1008662
470162-718EA 764.29 USD InStock
470162-718 470162-698
Torsion Inertia Device
Educational Materials Physics Educational Materials Mechanics Learning Activities
Experiment topics:
• Rotary oscillations
• Determination of moments of inertia using the oscillation method
• Moments of inertia of various geometric bodies
• Steiner's theorem

Axle for investigating rotational oscillation of various test bodies and for determining their moments of inertia from the period of oscillation. With ball-bearing mounted shaft, coil spring, and holding lug. The test body is a bar with weights that can be moved along its length and a circular disc with one hole in the center and eight away from the center for determining moments of inertia for eccentric axes of rotation and confirming Steiner’s theorem.

Deflecting torque of the spring: 0.028 Nm/rad.
Height of the torsional axle: approx. 200 mm
Cross bar:
Length: 620 mm
Weight: 135 g
Weights: 260 g each
Disc:
Diameter: 320 mm
Weight: 495 g
Boreholes: 8
Borehole spacing: 20 mm

Accessories for the torsion axle consists of two cylinders with nearly identical weights but different weight distributions, a mounting plate for the cylinders, a wooden disc, and a wooden sphere.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty