Would you like to log in again?
Electron Deflection Tube Demonstration Series
Supplier: AMERICAN 3B SCIENTIFIC SE
1000651
470162-714EA 1882.2 USD InStock
470162-714
Electron Deflection Tube Demonstration Series
Educational Materials Physics Educational Materials Modern Physics Learning Activities
Highly evacuated electron tube with focusing electron gun and fluorescent screen inclined relative to the beam axis, so that the path of the beam can be seen and the effects of electric and magnetic fields can be studied. The electron beam can be deflected electrically in the electric field of the built-in plate capacitor, and magnetically by using the Helmholtz coil pair (not included). By adjusting the electric field so that it cancels the magnetic deflection, it is possible to determine the specific charge (e/m) and the velocity of the electrons.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty