Would you like to log in again?
Electron Diffraction Tube Demonstration Series
Supplier: AMERICAN 3B SCIENTIFIC SE
1013885
470162-716EA 1843.59 USD InStock
470162-716
Electron Diffraction Tube Demonstration Series
Educational Materials Physics Educational Materials Modern Physics Learning Activities
High quality and reliable.
Highly evacuated electron tube for demonstrating the wave nature of electrons through the observation of interference caused by passage of electrons through a polycrystalline graphite lattice (Debye-Scherrer diffraction) and rendered visible a fluorescent screen. Also intended for determining the wavelength as a function of the anode voltage from the radii of the diffraction rings and the lattice plane spacing of graphite, as well as confirming de Broglie’s hypothesis. Required but not included is the tube holder and power supply.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty