Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

High Voltage Power Supply 5000 V

Supplier: AMERICAN 3B SCIENTIFIC SE
1003309
470162-726EA 960.49 USD InStock
470162-726
High Voltage Power Supply 5000 V
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Universally applicable, floating, high-voltage power supply with built-in, high voltage resistant transformer to supply the heater voltage for electron tubes. Continuously adjustable high voltage, safe to touch, with passive current limitation and analog voltage display. Unit poles can be connected to protected earth, but it is not center tapped.

Specifications:
High voltage output: 0-5000 V DC, max. 2 mA, max. 5 W
Heater voltage output: 6.3 V AC, max. 3 A
High voltage resistant up to 6 kV
Overload protection: Primary: fuse
Secondary: current-limiting resistors
Connections: 4 mm safety sockets
High-voltage display: Analogue
Dimensions: 235x130x155 mm³ approx.
Weight: 3.5 kg approx.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty