Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Millisecond Counter

Supplier: AMERICAN 3B SCIENTIFIC SE
1012833
470162-774EA 252.09 USD InStock
470162-774
Millisecond Counter
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Inexpensive, compact counter for measuring milliseconds (in conjunction with the free-fall apparatus 470162-756 ). Each count is started and stopped by a signal at the input sockets. The device is automatically reset to zero each time it is restarted. Includes plug-in power supply.

Time measurement: 1 ms – 9999 s
Supply voltage: 5 V DC
Connectors: 4-mm safety sockets
Dimensions: 105x75x35 mm approx.
Weight: 400 g approx.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty