Would you like to log in again?
Laser Reflection Sensor
Supplier: AMERICAN 3B SCIENTIFIC SE
1001034
470162-778EA 342.39 USD InStock
470162-778
Laser Reflection Sensor
Educational Materials Math Educational Materials Measurement Learning Activities
Sensor for triggering the digital counter in time measurements on moving objects. Suitable for opto electronic scanning of light and dark markings on moving objects or in conjunction with a reflecting foil to form a wide spaced obstruction sensor. The intensity of the laser beam is adjusted automatically according to the distance of the object. Includes reflecting foil, stand rod with screw thread, and connector lead with 8 pin miniDIN plugs.
Maximum range: 2.5 m
Laser power: <1 mW
Laser protection class: II
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty