Would you like to log in again?
Thermochemistry and Hess's Law
Supplier: ALDON CORP SE
IS8001
470163-068KT 53.65 USD InStock
470163-068
Thermochemistry and Hess's Law
Educational Materials Chemistry Educational Materials Thermochemistry/Kinetics Learning Activities
Hess' Law In Action!
  • Classic AP Chemistry Experiment
  • Instructions Included
  • Completed in Approximately 1 Hour
Employ Hess’s Law of enthalpy based on the observed data for two reactions. Students can measure the temperatures of the reaction in a calorimeter and analyze the data calculating the enthalpy of each reaction. 
Ordering information: Kit includes Hydrochloric Acid, Sodium Hydroxide, Ammonium Chloride, and Ammonium Hydroxide.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty