Would you like to log in again?
Activity Series Kit
Supplier: ALDON CORP SE
IS8002
470163-070KT 119.3 USD InStock
470163-070
Activity Series Kit
Educational Materials Chemistry Educational Materials Mixtures and Solutions Learning Activities
Oxidation and Reduction of Metals
  • Classic AP Chemistry Experiment
  • Determine Which Metal is the Most Reactive
  • Instructions Included
An activity series of metals is a table of metals arranged in the order of their decreasing chemical activity or the ease at which the metal will give up one or more electrons to form positive ions. This table is similar to the electrochemical series of elements. For example if you take the group of metals magnesium, mercury and nickel, magnesium is the most reactive and mercury the least. To empirically determine which of these metals is more reactive, place a piece of the metal in a salt solution of the other. The more reactive metal will replace the less reactive metal and the less reactive will appear in the solid form. The reactive metal has been oxidized; the less reactive metal has been reduced.
Ordering information: Includes Copper Metal, Zinc Metal, Magnesium Metal, Lead Metal, Copper Nitrate Solution, Zinc Nitrate Solution, Magnesium Nitrate Solution, Lead Nitrate Solution, Potassium Bromide, Potassium Bromate, Sulfuric Acid Solution, Chlorine Water, Iodine Water, Sodium Bromide Solution, Potassium Iodide Solution, Sodium Chloride Solution, Mineral Oil, 1 Empty Bottle (for Bromine Water), Teacher's Guide and Student Copymasters.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty