Would you like to log in again?
Ester Formation Kit
Supplier: ALDON CORP SE
IS8005
470163-076KT 65.05 USD InStock
470163-076
Ester Formation Kit
Educational Materials Chemistry Educational Materials Acids and Bases Learning Activities
Identify the Chemical by its SMELL!
  • Classic AP Chemistry Experiment
  • Instructions Included
  • Form various esters with this hands on experiment
The yield of ester can be increased either by removing one of the products of the reaction as it is formed or by increasing the concentration of one of the reactants. The normal procedure is to remove water using the dehydrating agent sulfuric acid, which also acts as the catalyst. In this experiment, we will combine various alcohols with acetic and butyric acids to form esters. The esters can be identified by their distinctive odors.
Includes:
Acetic Acid
n-Butyric Acid
n-Butyl Alcohol
Octyl Alcohol
Methyl Alcohol
Ethyl Alcohol
Sulfuric Acid
Glass Droppers
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty