Would you like to log in again?
Reaction Kinetics
Supplier: ALDON CORP SE
IS8007
470163-080KT 54.2 USD InStock
470163-080
Reaction Kinetics
Educational Materials Chemistry Educational Materials Thermochemistry/Kinetics Learning Activities
A Rate Law Experiment
  • Classic AP Chemistry Experiment
  • Vary Concentrations to change rate
  • Instructions Included
In a chemical reaction, reactants are converted to product at a given rate. This rate can be changed by alternating either the temperature of the reaction, the form of the reactants, the concentration of reactants or products or by adding a catalyst. The time it takes for a reaction to occur is monitored by observing a color change. By varying concentration of reactants the rate law is determined. By varying the temperature the activation energy is determined. The data should be graphed and analyzed during the lab so additional measurements can be made if necessary.
Ordering information: Includes Potassium Iodide Solution, Sodium Thiosulfate Solution, Hydrogen Peroxide Solution, Starch Soluble, Hydrochloric Acid Solution, and Pipettes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty