Would you like to log in again?
Dissociation Constants of Weak Acids Kit
Supplier: ALDON CORP SE
IS8008
470163-082KT 50.8 USD InStock
470163-082
Dissociation Constants of Weak Acids Kit
Educational Materials Chemistry Educational Materials Acids and Bases Learning Activities
Its All About Equilibrium!
  • Classic AP Chemistry Experiment
  • Instructions Included
Students determine the equilibrium constant of three unknown acids and compare the data to known values to identify them.
Ordering information: Includes Benzoic Acid Crystals, Potassium Hydrogen Phthalate, Sodium Bisulfate, Sodium Hydroxide, and Phenolphthalein Solution.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty