Would you like to log in again?
Quantitative Analysis
Supplier: ALDON CORP SE
IS8009
470163-084KT 97.25 USD InStock
470163-084
Quantitative Analysis
Educational Materials Chemistry Educational Materials Thermochemistry/Kinetics Learning Activities
Analytical Chemistry for AP Students
  • Classic AP Chemistry Experiment
  • Instructions Included
Colorimetric or turbidimetric methods of analysis have been developed for most of the elements and ionic species. These methods can be employed to quickly determine the chemical composition of an unknown substance. Before attempting to analyze an unknown it is useful to develop a logical process to follow. Care must be taken in sampling, collecting and washing precipitates, performing confirmatory tests and most importantly, recording data. It is also good laboratory practice to always analyze a known with each unknown as a double check on the reagents being used. The unknowns will be analyzed for one of six possible anions: Cl-, Br-, I-, PO42-, NO3-, and SO42-. A standard solution for each anion will also be run.
Includes:
Sodium Bromide Solution
Sodium Chloride Solution
Sodium Iodide Solution
Sodium Nitrate Solution
Sodium Phosphate Solution
Sodium Sulfate Solution
Ammonium Hydroxide
Mineral Oil
Ammonium Molybdate Solution
Diphenylamine
Barium Chloride Solution
Potassium Permanganate Solution
Ferric Nitrate Solution
Silver Nitrate Solution
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty