Would you like to log in again?
Beer Lambert Law
Supplier: ALDON CORP SE
IS8010
470163-086KT 42.95 USD InStock
470163-086
Beer Lambert Law
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Determine Concentrations of Unknowns
  • Classic AP Chemistry Experiment
  • Instructions Included
The probability that a photon of light will be absorbed by a solution is directly proportional to the concentration of the absorbing molecules, the thickness of the sample and the degree to which the molecules absorbs light. This relationship is called the Beer-Lambert Law and is used in analytical chemistry to determine concentrations of various compounds spectrophotometrically. A series of 5 solutions with different concentrations of iron are prepared and analyzed using the spectrophotometric assay for iron. 
Included:
Ferric Chloride Solution
Ascorbic Acid
1,10 Phenanthroline
Acetate Buffer
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty