Would you like to log in again?
Stoichiometry In The Synthesis Of An Ionic Compound
Supplier: ALDON CORP SE
IS8011
470163-088KT 53.05 USD InStock
470163-088
Stoichiometry In The Synthesis Of An Ionic Compound
Educational Materials Chemistry Educational Materials Ionic Properties Learning Activities
Create Potassium Alum Using Chemical Reactions
  • Classic AP Chemistry Experiment
  • Instructions Included
Many combinations of mono and trivalent cations yield crystals of the same stoichiometry and structure. The crystals are normally in the form of an octahedron. Sodium, potassium and ammonium ions are often the monovalent species whereas aluminum and chromium are examples of the trivalent ions. Potassium alum, KAl(SO4)2•12H2O, is the most common and is used in water purification, paper manufacturing and as a mordant in dyeing.  In this experiment we will perform a series of chemical reactions which lead to the synthesis of the ionic compound potassium alum.
Includes:
Aluminum Metal Powder
Potassium Hydroxide Solution
Sulfuric Acid Solution
Ethanol/Water 50:50 V/V Solution
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty