Would you like to log in again?
Equilibrium Constant Of An Ionic Compound AP Chemistry Kit
Supplier: ALDON CORP SE
IS8012
470163-090KT 33 USD InStock
470163-090
Equilibrium Constant Of An Ionic Compound AP Chemistry Kit
Educational Materials Chemistry Educational Materials Ionic Properties Learning Activities
Determine The Equilibrium Constant for Colored Species
  • Classic AP Chemistry Experiment
  • Instructions Included
The idea of the reversibility of chemical reactions was first stated by C. Berthollet in 1799. He noted deposits of sodium carbonate in certain salt lakes in Egypt and concluded that they were produced by the high concentration of sodium chloride and dissolved calcium carbonate. This is the reverse of the standard laboratory procedure in which calcium carbonate is produced from the reaction on sodium carbonate and calcium chloride. A standard curve will be established for the absorbance of a colored species and then used to determine the concentrations of unknown solutions. The results will allow us to determine the equilibrium constant for the formation of the colored species.
Ordering information: Includes Iron(III) Nitrate Solution, Potassium Thiocyanate Solution, and Nitric Acid Solution.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty