Would you like to log in again?
Determination of the Molecular Mass of a Liquid
Supplier: ALDON CORP SE
IS8018
470163-102KT 48.95 USD InStock
470163-102
Determination of the Molecular Mass of a Liquid
Educational Materials Chemistry Educational Materials Properties of Matter Learning Activities in Chemistry
Simple, Hands On Experiment
  • Classic AP Chemistry Experiment
  • Instructions Included
A volatile liquid is placed in a test tube and the tube is closed with a stopper with a hole in it. The test tube is placed in a hot water bath. The liquid vaporizes and excess vapors escape through the hole. The tube is then placed in an ice bath to quickly cool the vapor and cause it to condense. The mass of the tube is determined.
Includes:
Methyl Alcohol
Isopropyl Alcohol
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty