Would you like to log in again?
Thermochromism
Supplier: ALDON CORP SE
IS8019
470163-104KT 126.95 USD InStock
470163-104
Thermochromism
Educational Materials Chemistry Educational Materials Properties of Matter Learning Activities in Chemistry
How Structure Changes Color
  • Classic AP Chemistry Experiment
  • Instructions Included
The way in which light is absorbed or reflected by a crystalline material is determined by the structure of the crystal. This crystal structure can be changed by the application of heat. In some cases this change is reversible and the original structure, and therefore the original color, returns upon cooling. Two compounds are formed when metal salts are reacted with the [HgI4]2- Ion. Each compound is heated on a hot plate and a change in color is noted. The initial color of the compound returns upon cooling.
Includes:
Mercury(II) Chloride
Potassium Iodide
Silver Nitrate
Copper(II) Chloride, anhydrous
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty