Would you like to log in again?
Determination of the Properties of Buffer Solutions
Supplier: ALDON CORP SE
IS8024
470163-112KT 75.2 USD InStock
470163-112
Determination of the Properties of Buffer Solutions
Educational Materials Chemistry Educational Materials Acids and Bases Learning Activities
What Does A Buffer Do?
  • Classic AP Chemistry Experiment
  • Instructions Included
In this experiment, you will prepare three buffer solutions having different pH values and show that the pH of these solutions does not change significantly when small amounts of acids and bases are added. You will also show that when the same amounts of acids and bases are added to water and to a non-buffer solution (e.g. NaCl solution), the pH changes are large.
Includes:
Sodium Acetate, trihydrate
Ammonium Chloride
Sodium Bicarbonate
Sodium Chloride
Acetic Acid EZ-Prep
Ammonium Hydroxide EZ-Prep
Sodium Hydroxide Solution
Hydrochloric Acid Solution
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty