Would you like to log in again?
Determination of the Empirical Formula of Magnesium Oxide
Supplier: ALDON CORP SE
IS8025
470163-114KT 31.4 USD InStock
470163-114
Determination of the Empirical Formula of Magnesium Oxide
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Quantitative Stoichiometry
  • Classic AP Chemistry Experiment
  • Instructions Included
The quantitative stoichiometric relationships governing mass and amount will be studied using the combustion reaction of magnesium metal. Magnesium is reacted with oxygen from the air in a crucible, and the mass before and after the oxidation is measured. The resulting masses are used to calculate the experimental empirical formula of magnesium oxide, which is then compared to the theoretical empirical formula. A crucible and Bunsen burner will be used to heat magnesium metal to burning. This lab illustrates the (i) law of conservation of mass and (ii) the law of constant composition.
Includes:
Magnesium Metal
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty