Would you like to log in again?
Acid-Base Indicators
Supplier: ALDON CORP SE
IS8027
470163-118KT 59.15 USD InStock
470163-118
Acid-Base Indicators
Educational Materials Chemistry Educational Materials Acids and Bases Learning Activities
How Do Indicators Work?
  • Classic AP Chemistry Experiment
  • Instructions Included
pH is the measure of the concentration of hydrogen ions in a solution. As this concentration can extend over several orders of magnitude, it is convenient to express it by means of logarithms of base ten. Certain pigments change color with a change in pH. In this experiment we will extract pigments from various sources and determine if they are sensitive to changes in pH.
Ordering information: Includes Hydrochloric Acid, Sodium Hydroxide, Isopropyl Alcohol, and Acetone.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty