Would you like to log in again?
Gravimetric Analysis
Supplier: ALDON CORP SE
IS8028
470163-120KT 43 USD InStock
470163-120
Gravimetric Analysis
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Determine the Unknown!
  • Classic AP Chemistry Experiment
  • Instructions Included
Determine the identity of a metal carbonate by gravimetric analysis. Gravimetric analysis is a technique through which the amount of an analyte (the ion being analyzed) can be determined through the measurement of mass. You will complete the analyses comparing the masses of two compounds containing the analyte. The formula weight and the identity of the unknown are determined using gravimetric analysis.
Includes:
Calcium Chloride, anhydrous
Silver Nitrate
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty