Would you like to log in again?
Analysis of Potassium Aluminum Sulfate 12-Hydrate
Supplier: ALDON CORP SE
IS8029
470163-122KT 30.4 USD InStock
470163-122
Analysis of Potassium Aluminum Sulfate 12-Hydrate
Educational Materials Chemistry Educational Materials Gas Analysis Learning Activities
Determine if you made the correct compound
  • Classic AP Chemistry Experiment
  • Instructions Included
After a compound has been synthesized, analytical tests should be carried out to confirm that the correct compound has been produced. In this lab we will perform tests to confirm that the crystals formed are in fact potassium aluminum sulfate 12 hydrate by comparing the melting point to published results, finding the number of waters of hydration, and determining the percent of sulfate in the compound.
Includes:
Aluminum Potassium Sulfate
Barium Nitrate
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty