Would you like to log in again?
Liquid (Column) Chromatography
Supplier: ALDON CORP SE
IS8031
470163-126KT 84.05 USD InStock
470163-126
Liquid (Column) Chromatography
Educational Materials Chemistry Educational Materials Chromatography Learning Activities
How Do Compounds Exit the Column?
  • Classic AP Chemistry Experiment
  • Instructions Included
The mixture to be analyzed by column chromatography is applied to the top of the column. The liquid solvent (the eluent) is passed through the column by gravity or by the application of air pressure. Equilibrium is established between the solute adsorbed on the adsorbent and the eluting solvent flowing down through the column. Because the different components in the mixture have different interactions with the stationary and mobile phases, they will be carried along with the mobile phase to varying degrees and a separation will be achieved. The individual components, or elutants, are collected as the solvent drips from the bottom of the column.
Ordering information: Kit includes Isopropyl Alcohol, Silica Gel, Dye Mixture, Pipettes, and Glass Wool.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty