Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Viking Rocket

Supplier: ESTES INDUSTRIES INC SE
1949
470163-206CS 449.25 USD InStock
470163-206
Viking Rocket
Educational Materials Physics Educational Materials Applied Physics Learning Activities
The perfect beginner Rocket

  • Length: 12.1" (30.7 cm)
  • Diameter: .74" (19 mm)
  • Estimated Weight: .6 oz. (17 g)
  • Projected Max. Altitude: 1600 ft. (488 m)
  • Skill Level 1

The Viking can be built with three, four or five fins in 48 possible arrangements. This makes it perfect to use for comparison studies in aerodynamics. In fact, the Viking is so popular there’s even a GEMS book written for it. No matter how you place the fins, this unique, adjustable rocket offers the ultimate in high performance model rocket flight!

This easy-to-build, Skill Level 1 rocket kit includes a molded nose cone, a single piece body tube, card stock fins and self-stick decals. It’s another friction fit, standard engine rocket that requires simple painting. The Viking can reach amazing launch heights over 1,600 feet and will drift back to earth on a bright streamer. Skill Level 1. Laser cut Card Stock Fins, Self-stick Decals, Streamer Recovery.

Ordering information: Recommended Engines 1/2A6-2, A8-3, A8-5, B4-4, B6-4, B6-6, C6-5, C6-7. Recovery wadding required but not included.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty