Would you like to log in again?
Separation of a Mixture: Physical Properties of Matter
Supplier: ALDON CORP SE
IS1000
470163-288KT 61.9 USD InStock
470163-288
Separation of a Mixture: Physical Properties of Matter
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Perfect Introduction to Physical Properties.
  • Ideal for beginner scientists
  • Separate a mixture based on only physical characteristics
  • All chemicals required are included
After a class discussion of matter and the fact that matter may differ from other matter in a variety of ways, students will examine three different solids individually and observe some differences in physical properties. Sand, iron filings and salt will be observed and tested using water and magnets. Based on what they find individually they will come up with a method (as a class or as groups) to separate a mixture of all three.
Ordering information: Includes Sand, Salt, Iron Filings, 3 Part Mixture, Magnets, Plastic Cups, Magnifiers, and Teacher's Guide and Student Copymaster.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty